Learn More
Proteintech Group Inc TurboGFP-Trap Agarose beads, kit
Supplier: Proteintech Group Inc TBTAK20
TurboGFP-Trap Agarose is an affinity bead for immunoprecipitation (IP) of TurboGFP fusion proteins It comprises an anti-TurboGFP VHH/ Nanobody coupled to agarose beads.
This item is not returnable.
View return policy