Learn More
Chemglass Life Sciences Tubing, Quartz, 28mm ID x 31mm OD, 4ft Length
Supplier: Chemglass Life Sciences CGQ0800T90
Tubing, Quartz, 28mm ID x 31mm OD, 4ft Length. General Electric Type 214 grade clear fused quartz tubing. Tubing is stocked in 4ft lengths. PLEASE NOTE: PRICE LISTED IS PER LENGTH OF 4ft TUBING.