Learn More
Chemglass Life Sciences FEP Tubing; 3/8" OD x 1/4" ID x 10ft Long
Supplier: Chemglass Life Sciences CG197510
FEP Tubing; 3/8" OD x 1/4" ID x 10ft Long. EP heavy wall tubing for use with Chemglass process reactors. Used to connect components having 1/4", 1/2", or 3/4"OD glass connections.