Learn More
Saint Gobain Performance Plastics Tube Tygon 5/32 x 9/32 50FT/PK
Supplier:  Saint Gobain Performance Plastics ACF00010
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Diameter Inner 5/32 in. Diameter Outer 9/32 in. Maximum Pressure 30 psi at 73 deg.F Thickness 1/16 in. Clear
Catalog No. 50-206-8940
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.