Learn More
Saint Gobain Performance Plastics Tube Tygon 5/32 x 7/32 50FT/PK
Supplier:  Saint Gobain Performance Plastics ACF00009
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Diameter Inner 5/32 in. 4 mm Diameter Outer 7/32 in. 5.6 mm Maximum Pressure 17 psi at 73 deg.F Thickness 1/32 in. 0.8 mm Overall Length 15 m Clear Must order minimum quantity of 2.
Catalog No. 50-206-8894