Learn More
Andwin Scientific TUBE TABS 100/BAG
Supplier: Andwin Scientific C32712
The GeneMate Tube Tabs are ideal for extracting tubes from centrifuges, heatblocks, boiling water, etc. The tube tabs are autoclavable. This product is sold in a bag of 100.