Learn More
Electron Microscopy Sciences Tube Rocker, 115V, 13.3 x 5.3 x 4"
Supplier: Electron Microscopy Sciences 6570011
The TubeRocker is designed to provide the ideal speed and tilt angle for the agitation of sample tubes. The grooved design of the included rubber mat properly secures all tube sizes ranging from 1.5 to 50 ml, without the need for added straps or bands. Small 5 x 13" size makes it easy to fit on any lab bench. It can be conveniently placed inside refrigerators, cold rooms or laboratory incubators. Simple-to-use. Needs no adapters or mounting hardware. Just press the power switch and let it rock!
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.