Learn More
Ace Glass, Inc. TUBE 35ML 150PSI T-WELL C
SDP
Supplier:  Ace Glass, Inc. 8648170
Encompass_Preferred
35mL pressure tube with #15 Ace-Thred, rated for 150psig at as much as 120°C. Includes #15 front-seal plunger valve with Thermowell. Dimensions: 25.4mm O.D. x 17.8cm long.
Catalog No. 50-976-211
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.