Learn More
MTC Bio Test Tube / Culture Tube, 13x100mm, 8mL, PS, rimless, no cap, non-sterile, 10 bags of 300 tubes, 3000/cs
Supplier:  MTC Bio T1411
Encompass_Preferred
MTC Bio’s test tubes are packaged for high volume users in bulk quantity economy cases. Supplied non-sterile and without caps (available separately), all three sizes are available in translucent polypropylene or crystal clear polystyrene. Plug-type caps are available separately. RNase, DNase, pyrogen, & endotoxin free.
Catalog No. 50-226-8497
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.