Learn More
Andwin Scientific TUBE 12X75MM, NS, 5ML, CS/1000
Supplier: Andwin Scientific 60818430
Polypropylene Culture Tube Without caps. Autoclavable. Size mm: 12x75. Volume: 5mL. Bulk packaging. Endotoxin-free.