Learn More
MTC Bio Transport/ Culture Tube, 16 x1 00mm, PS, round bottom, with screw-cap, white printed graduations, 12mL, sterile, 10 bags of 100 tubes, 1000/cs
Supplier:  MTC Bio T7955
Encompass_Preferred
These 16 x 100 mm screw cap test tubes are available in transparent polypropylene with a skirted self-standing bottom or in crystal clear polystyrene with a traditional round bottom. Each tube has easy to read graduations and a frosted white marking area (12x50mm). Packed 100 tubes (with attached caps) per sterile bag, 10 bags per unit. RNase, DNase, pyrogen, & endotoxin free.
Catalog No. 50-226-8501
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.