Learn More
Saint Gobain Performance Plastics TUB TYG 2475 5/16 x 7/16 50FT/PK
Supplier:  Saint Gobain Performance Plastics ACG00022
Encompass_Preferred
Saint-Gobain Tygon Formula 2475 High-Purity Tubing Diameter Inner 5/16 in. 7.9 mm Diameter Outer 0.43 in. 11 mm Thickness 1.6 mm 0.06 in. Certifications/Compliance Meets USP Class VI FDA and NSF specifications. Documented biocompatibility to ISO 10993 standards.
Catalog No. 50-206-8979
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.