Learn More
Saint Gobain Performance Plastics TUB TYG 2475 1/4 x 3/8 50FT/PK
Saint-Gobain Tygon Formula 2475 High-Purity Tubing Diameter Inner 1/4 in. 6.4 mm Diameter Outer 0.37 in. 9.5 mm Thickness 1.6 mm 0.06 in. Certifications/Compliance Meets USP Class VI FDA and NSF specifications. Documented biocompatibility to ISO 10993 standards.