Learn More
Electron Microscopy Sciences T-Sue™ Microarray Mold Kit, 2 mm
Supplier:  Electron Microscopy Sciences 6913202
Encompass_Preferred
T-Sue array molds are specifically designed to be simple, easy to use and inexpensive. The tissues can be analyzed in consistent conditions enhancing the efficiency of the research. Allow you to perform TMAs more quickly with excellent results. By using array molds, you can process up to 170 specimens onto one single slide in very little time.
Made of silicone, these molds can withstand temperatures between -100 deg C to +250 deg C and be reused hundreds of times without losing their flexibility.
Each kit includes one T-Sue array mold and four punch needles with stylet.
Size: 2 mm
Capacity: 24 cores
Catalog No. 50-193-0033
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.