Learn More
TSLP Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Pepro Tech Inc 500P2581MG
Description
AA Sequence of recombinant protein: MYDFTNCDFE KIKAAYLSTI SKDLITYMSG TKSTEFNNTV SCSNRPHCLT EIQSLTFNPTAGCASLAKEM FAMKTKAALA IWCPGYSETQ INATQAMKKR RKRKVTTNKC LEQVSQLQGLWRRFNRPLLK QQ. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human TSLP. Anti-Human TSLP-specific antibody was purified by affinity chromatography employing an immobilized Human TSLP matrix. Sandwich ELISA: To detect Human TSLP by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human TSLP (500-P258Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hHuman TSLP. Western Blot: To detect Human TSLP by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human TSLP is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.Specifications
TSLP | |
Polyclonal | |
Unconjugated | |
TSLP | |
Thymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
85480 | |
-20°C |
ELISA, Western Blot | |
0.1-1.0 mg/mL | |
PBS with no preservative | |
Q969D9 | |
TSLP | |
E.coli-derived Recombinant Human TSLP | |
1 mg | |
Primary | |
Human | |
Lyophilized |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.