Learn More
ACROBiosystems ACROBiosystems Cynomolgus TSLP Protein, His Tag
Supplier:  ACROBiosystems TSPC52H81MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus TSLP, His Tag (TSP-C52H8) is expressed from human 293 cells (HEK293). It contains AA Tyr 29 - Gln 159 (Accession # A0A2K5TXV0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 17.1 kDa.
Catalog No. 50-195-6395
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.