Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TSKgel™ ODS-100Z Guard Cartridge, Sold by MilliporeSigma™ Supelco™
The workhorse for analysis of small molecular weight compounds in life science applications
Supplier: MilliporeSigma Supelco 821995
Description
- The bonded phase of TSKgel ODS-100Z columns is prepared by reacting the silica surface with a difunctional octadecylsilane reagent, followed by repeated endcapping with a monofunctional trimethylsilane reagent.
- The resulting TSKgel ODS-100Z columns contain a high density monomeric C18 bonded phase for maximum retention and selectivity of small molecular weight compounds and minimal secondary interaction with residual silanol groups.
- This general purpose reversed phase column is the workhorse for analysis of small molecular weight compounds in life science applications.
- Lot-to-lot reproducibility stands out.
- Shipped in acetonitrile.
- 20% Carbon, encapped with trimethylsilane
- 0-100% organic solvent
Specifications
5 μm | |
450 m2/g | |
L1 Per USP | |
10°C to 50°C | |
Reversed phase |
1 cm | |
2 mm | |
C18 (octadecyl) | |
Stainless steel | |
Spherical silica |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TSKgel is a registered trademark of Tosoh Corporation
Spot an opportunity for improvement?Share a Content Correction