Learn More
Thermo Scientific™ TSG Series Undercounter Refrigerators
Benefit from innovative refrigeration solutions for healthcare and research needs
Supplier: Thermo Scientific™ TSG505SD
Description
Powered by Phononic solid state refrigeration technology, The Thermo Scientific™ TSG Series Undercounter Refrigerators have been designed to meet the rigorous and demanding needs of healthcare, research and industrial applications where solid performance is required, in support of breastmilk and medication storage, analyzer kits and reagents, and even cell culture media.
Key features of the TSG505 Undercounter Refrigerator:
- Solid state refrigeration technology
- Whisper quiet operation leading to a more comfortable work environment supporting clinical and healthcare needs
- Increased capacities due to fewer refrigeration components means you can store more in less space
- Energy Star rated performance meaning lower operating costs
- Green refrigerants supporting global initiatives to reduce green house gas emissions
- Full alarm set, including hi/lo temperature, door ajar and power fail to support requirements for medical storage
- Remote alarm contacts on TSG505, supporting requirements of research for monitoring
Warranty: varies by country, please contact us for details
Specifications
2°C to 9°C | |
5.5 cu. ft. | |
100 - 240 V 50-60 Hz | |
NEMA 6-15 P | |
CRS Powder Coated | |
No | |
26.9 in. (door closed) | |
Energy efficient | |
1 | |
Undercounter |
CE, UL | |
156 L | |
100/240 V | |
Remote Alarm Contacts | |
Right | |
23 in. | |
684 mm | |
Alarm | |
3 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.