Learn More
Thermo Scientific™
TSG Series General Purpose Chromatography Refrigerators
Designed for your everyday laboratory needs
Supplier: Thermo Scientific™ TSG49CSGA
Description
Thermo Scientific™ TSG Series general-purpose chromatography refrigerators are designed with your needs in mind. The TSG Series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing non-critical products such as reagents, media, proteins, and other samples requiring a 2°to 10°C controlled environment.
• Hydrocarbon safety sensor ensures cargo and lab personnel are kept safe
• Featuring a wide range of cabinet sizes from 30 cu. ft. to 72 cu. ft.
• Whisper-quiet operation
• ENERGY STAR certified
• Temperature, door ajar*, and power failure (with battery back-up) alarms
• Green, natural refrigerants
• Adaptive defrost
• Glass (double pane) or solid door configurations
• Choice of white painted interior and exterior, or stainless steel interior and gray exterior
• Digital controller
• Rear access port
• Remote alarm contacts
• Keyed door lock(s)
• LED interior lighting
• Low heat output
• 2 years parts and labor, plus 3 year compressor warranty
*All models except TSG45CPLA and TSG45CSLA come equipped with door ajar alarms
Specifications
TSG Series general purpose chromatography refrigerator | |
1387 L | |
+5°C | |
Glass | |
8 Full, 2 Half | |
45 kg/full shelf, 22.5 kg/half shelf | |
Painted | |
cULus, ENERGYSTAR, Prop 65 | |
1 Rear access port and 2 side access ports | |
Adaptive Defrost | |
Key Lock | |
32 x 52.7 x 81 in.(813 x 1339 x 813 mm) | |
NEMA 5-15 P | |
115 V 60 Hz | |
Energy efficient | |
271 kg |
49 cu. ft. | |
2°C to 10°C | |
R290 | |
2 | |
100 lb./full shelf; 50 lb./half shelf | |
Gray-painted | |
Stainless Steel | |
Digital Microprocessor | |
2 years parts and labor, plus 3-year compressor warranty | |
Alarm | |
28.5 x 49 x 61.75 in. (724 x 1245 x 1569 mm) | |
8.8 A | |
115 V | |
60 Hz | |
597 lb. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.