Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrkA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA595139
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL.
NTRK1 (TRKA) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. TrkA is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of TrkA leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in TrkA have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer.Specifications
TrkA | |
Polyclonal | |
Unconjugated | |
Ntrk1 | |
C80751, DKFZp781I14186, EC 2.7.10.1, gp140trk, high affinity nerve growth factor receptor, kinase TrkA, MTC, neurotrophic receptor tyrosine kinase 1, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, Ntrk1, Oncogene TRK, p140-TrkA, Slow nerve growth factor receptor, Tkr, TRK, TRK1, TRK1-transforming tyrosine kinase protein, Trka, Trk-A, TrkA neurotrophin receptor, trkA proto-oncogene receptor, tropomyosin-related kinase A, Tyrosine kinase receptor, Tyrosine kinase receptor A | |
Rabbit | |
Affinity chromatography | |
RUO | |
18211, 59109 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5MG BSA and 0.05MG sodium azide, 0.05MG thimerosal | |
P35739, Q3UFB7 | |
Ntrk1 | |
A synthetic peptide corresponding to a sequence at the N-terminus of mouse TrkA (71-90aa LYVENQQHLQRLEFEDLQGL). | |
100 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction