Learn More
World Precision Instrument Trio 3-Axis Micromanipulator-SU-TRIO235
SDP
Supplier:  World Precision Instrument SUTRIO235
Encompass_Preferred
The new SU-TRIO245 is a highly stable 3-axis manipulator with 25 mm of travel on each axis. The TRIO's synthetic 4th axis can be set in software as any angle between 0 and 90º for diagonal movement. Based on a lead-screw design with a smaller overall size and footprint than most manipulators, the SU-TRIO245 is ideal for applications that require 2 pipettes in one setup or for setups where space is limited. 
Catalog No. 50-253-8510
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.