Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl Citrate, FCC, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18607570
Click to view available options
Quantity:
500 mL
4 L
5 gal.
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Pail

T1171, 77-93-0, C12H20O7

Spectrum™ Chemical Triethyl Citrate, FCC is a colorless, odorless liquid used as a food additive (E number E1505) to stabilize foams, especially as whipping aid for egg whites. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplements and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 77-93-0
Molecular Formula C12H20O7
Molecular Weight (g/mol) 276.29
InChI Key DOOTYTYQINUNNV-UHFFFAOYSA-N
IUPAC Name 1,2,3-triethyl 2-hydroxypropane-1,2,3-tricarboxylate
SMILES CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC

Specifications

CAS Max % 1
Packaging Poly Pail
Refractive Index 1.439-1.443
Quantity 5 gal.
Percent Purity 99.0 to 100.5%
Grade FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.