Learn More
Enzo Life Sciences Triacetyl resveratrol (10mg). CAS: 42206-94-0
SDP
Supplier:  Enzo Life Sciences BMLFR1190010
Encompass_Preferred
A resveratrol prodrug. One approach to increasing the half life of resveratrol in vivo is acetylation of the phenolic OH groups. Deacetylation presumably takes place in vivo or in intact cells via the action of intracellular esterases. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (warm) (8mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0839
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.