Learn More
Cole-Parmer Ashcroft IXLDP50IWC Low Diff Pressure Transmitter, 50.0" WC, 4-20mA, 0.5%
Supplier:  Cole-Parmer 6843942
Encompass_Preferred
Designed for the measurement and control of very low pressure and flow in industrial and process plant environments. The transmitter features s rugged NEMA4X enclosure, built-in electrical terminal box isolated from the electronics and threaded process connections.
Catalog No. 50-992-719
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.