Learn More
Biotang Inc Transformation buffer-2 (10 mM MOPS 75 mM Calcium Chloride 10 mM KCL 15% Glycerol pH 7.5), 125 ml
Supplier:  Biotang Inc BTBB586
Encompass_Preferred
Transformation buffer-2 (10 mM MOPS 75 mM Calcium Chloride 10 mM KCL 15% Glycerol pH 7.5), 125 ml
Catalog No. 50-152-105
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.