Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-(Boc-aminomethyl)cyclohexylamine, 97%, Thermo Scientific Chemicals

Catalog No. AAH62564MD
Click to view available options
Quantity:
250 mg
1 g
5 g

trans-4-(Boc-aminomethyl)cyclohexylamine a compound with potential for biochemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-(Boc-aminomethyl)cyclohexylamine a compound with potential for biochemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 192323-07-2
Molecular Formula C12H24N2O2
Molecular Weight (g/mol) 228.34
MDL Number MFCD06657670,MFCD12408630,MFCD22395596
InChI Key HMMYZMWDTDJTRR-UHFFFAOYSA-N
Synonym cis-4-boc-aminomethyl cyclohexylamine, tert-butyl trans-4-aminocyclohexylmethylcarbamate, tert-butyl trans-4-aminocyclohexyl methyl carbamate, tert-butyl cis-4-aminocyclohexyl methyl carbamate, trans-4-boc-aminomethyl cyclohexylamine, 4-boc-aminomethyl-cyclohexylamine, tert-butyl n-4-aminocyclohexyl methyl carbamate, tert-butyl 4-aminocyclohexyl methyl carbamate, tert-butyl trans-4-aminocyclohexyl methylcarbamate
PubChem CID 21882540
IUPAC Name tert-butyl N-[(4-aminocyclohexyl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1CCC(N)CC1

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 228.33
Percent Purity 97%
Chemical Name or Material trans-4-(Boc-aminomethyl)cyclohexylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.