Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-3-Hydroxycinnamic acid, 99%, Thermo Scientific Chemicals

Catalog No. AAA1446622
Click to view available options
Quantity:
10 g
50 g
100 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14755-02-3
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00004390
InChI Key KKSDGJDHHZEWEP-SNAWJCMRSA-N
Synonym 3-hydroxycinnamic acid, 3-coumaric acid, m-coumaric acid, m-hydroxycinnamic acid, 3-3-hydroxyphenyl acrylic acid, e-3-3-hydroxyphenyl acrylic acid, trans-3-hydroxycinnamic acid, trans-3-hydroxycinnamate, cinnamic acid, m-hydroxy, 2e-3-3-hydroxyphenyl prop-2-enoic acid
PubChem CID 637541
ChEBI CHEBI:32357
IUPAC Name (E)-3-(3-hydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=CC(=C1)O)C=CC(=O)O

Specifications

Melting Point 192°C to 196°C
Quantity 100 g
Beilstein 2690254
Formula Weight 164.16
Percent Purity 99%
Chemical Name or Material trans-3-Hydroxycinnamic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-615-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.