Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,3-Cyclohexanediamine 97.0+%, TCI America™
SDP

Catalog No. C2802200MG
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 26883-70-5
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
InChI Key GEQHKFFSPGPGLN-WDSKDSINSA-N
Synonym trans-1,3-Diaminocyclohexane
PubChem CID 6999482
IUPAC Name (1S,3S)-cyclohexane-1,3-diamine
SMILES C1CC(CC(C1)N)N

Specifications

Color Colorless
Boiling Point 198°C
Quantity 200 mg
UN Number 2735
Formula Weight 114.19
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material trans-1,3-Cyclohexanediamine
EINECSNumber (3)-3871
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.