Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,2-Bis(diphenylphosphino)ethylene, 98%, Thermo Scientific Chemicals

Catalog No. AAL0850806
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 983-81-3
Molecular Formula C26H22P2
Molecular Weight (g/mol) 396.41
MDL Number MFCD00003046 MFCD00063219
InChI Key NCKJIJSEWKIXAT-QURGRASLSA-N
Synonym trans-1,2-bis diphenylphosphino ethylene, e-1,2-bis diphenylphosphino ethene, ethene-1,2-diylbis diphenylphosphane, e-vinylenebis diphenylphosphine, cis-1,2-bis diphenylphosphino ethylene, trans-1,2-ethenediylbis diphenylphosphine, trans-vinylenebis diphenylphosphine, cis-vinylenebis diphenylphosphine
PubChem CID 2733288
IUPAC Name [(E)-2-diphenylphosphanylethenyl]-diphenylphosphane
SMILES C(=C/P(C1=CC=CC=C1)C1=CC=CC=C1)\P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 126°C to 130°C
Quantity 5 g
Beilstein 757793
Sensitivity Air Sensitive
Formula Weight 396.41
Percent Purity 98%
Chemical Name or Material trans-1,2-Bis(diphenylphosphino)ethylene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-570-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.