Learn More
Research Products International Corp Traceable Jumbo Digital Thermometer with Large Sensor
SDP
Supplier:  Research Products International Corp 814548
Encompass_Preferred
Ideal Digital Thermometer for monitoring temperatures in refrigerators, freezers, incubators, and water baths. Available with glass enclosed temperature buffered sensor that provides a stable reading when refrigerator or freezer door is opened. Also offered with a 3/16'' diameter x 3/4'' probe for immersion in tubes, heating blocks and water baths. Displays minimum, maximum, current probe temperature plus room temperature. Temperature range: -58? to 158?F (-50 to 70?C) with accuracy of +1?C. Additonal features include: built-in alarm and sealed probe. Supplied with battery, ceritificate of calibration and 10 foot cable.
Catalog No. 50-999-178
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.