Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Top Flasks
Neck Size: 24 mm
Supplier: MilliporeSigma™ Supelco™ 21001U
Specifications
24 mm | |
1/Pk. | |
Top Flask | |
300 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction