Learn More
Sino Biological TNFSF12 Protein 500ug (Sino Biological)
Supplier:  Sino Biological 90094C04H500
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
A DNA sequence encoding the human / cynomolgus TNFSF12 (F7HGN4) (Ser94-His249) was expressed with the Fc region of mouse IgG1 at the N-terminus. Human and Cynomolgus TNFSF12 sequences are identical.
Catalog No. 50-237-1839