Learn More
Sino Biological TNFSF12 Protein 50ug (Sino Biological)
Supplier:  Sino Biological 90094C04H50
Encompass_Preferred
A DNA sequence encoding the human / cynomolgus TNFSF12 (F7HGN4) (Ser94-His249) was expressed with the Fc region of mouse IgG1 at the N-terminus. Human and Cynomolgus TNFSF12 sequences are identical.
Catalog No. 50-237-1840
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.