Learn More
Enzo Life Sciences TNF-alpha (human), ELISA kit (5x96wells)
Supplier: Enzo Life Sciences ADI901099
Highly sensitive ELISA for measuring TNF-α, the primary mediator of immune regulation. The TNF-α (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4 hours. Absorbance is read at 450 nm. Assay up to 39 samples in duplicate per 96-well plate. The broad dynamic range makes assaying different pathologies easy. The larger package size offers a lower cost-per-test. Alternative name: Tumor necrosis factor-alpha, TNFSF 2. Sensitivity: 8.43 pg/ml (range 15.63 - 1,000 pg/ml). Use/Stability: Store all components at +4°C, except standard at -20°C. UniProt ID: P01375. Assay Time: 4 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human . Content: Microtiter plate, Conjugate, Antibody, Assay buffer 13 concentrate, Wash buffer concentrate, Standard, TMB Substrate, Stop solution 2.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.