Learn More
Enzo Life Sciences TN-16 (50 mg). CAS: 33016-12-5
SDP
Supplier:  Enzo Life Sciences BMLT1200050
Encompass_Preferred
A novel synthetic inhibitor of microtubule formation which binds to the colchicine site. It prevents the stabilization of microtubules in taxol-treated cells. Blocks the cell cycle in M-phase. Purity: ≥97% (HPLC). Long Term Storage: Ambient.
Catalog No. 50-201-2213
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.