Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences (TMS) Tetramethylsilane 100 ML

Supplier:  Electron Microscopy Sciences 21760

Encompass_Preferred

CAS #75-76-3 Si(CH₃)₄
Boiling point: 26.6-26.7°C

An organo-silicon compound, tetramethylsilane (TMS) is an ideal solvent for tissue drying for scanning electron microscopy. It is chemically inert, has as low boiling point of 26.6°C (lower than acetone, diethyl ether and 1,2-epoxy propane), and is soluble in most organic solvents but insoluble in water. These properties of TMS result in excellent tissue surface details.

For details please see Dey, Sudip. Et al. (1989). A new rapid method of air-drying for scanning electron microscopy using tetramethylsilane. J. of Microscopy, Vol 156, pp. 259-261.

Catalog No. 50-294-93


Explore all special offers
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.