Learn More
Neogen Diagnostics TMB Membrane Plus Substrate, 1 L, HRP, Membrane, Colorimetric
Supplier:  Neogen Diagnostics 337177
Encompass_Preferred
TMB Membrane Plus Substrate is a ready-to-use solution. It contains 3,3’,5,5’-tetramethyl-benzidine in a mildly acidic buffer in a one bottle format. TMB Membrane Plus Substrate develops an insoluble, permanent dark blue reaction in the presence of peroxidase. The substrate is intended for western blot, dot blots, and line assays and offers comparable performance and sensitivity to some chemiluminescent substrates. This substrate is not recommended for microwell or immunohistochemical applications. Stable for 2 years, from the date of manufacture, when stored at room temperature (15 to 25C) or 48 months when stored at 2-8C.
Catalog No. NC2215582
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.