Learn More
Hach Company EDTA Digital Titrator Cartridge, 0.800 M
Supplier: Hach Company 1439901
For determination of Calcium and Total Hardness by Digital Titrator titration. 0.800 M EDTA. Digits = mg/L CaCO3 in 100 mL sample. Approximately 40-100 tests.