Learn More
Hach Company AT1000 Series Potentiometric Titrator, 2 Burettes, 2 Pumps
Supplier:  Hach Company AT1222.97
Encompass_Preferred
The AT1222 is a reliable, accurate, and easy to use instrument that simplifies complex titrations in the laboratory. This titrator was designed in Germany as part of the Hach Titralab AT1000 family of automatic titrators and includes two burettes, two pumps and two measurement inputs. The titrator is delivered to you preassembled for easy installation and does not require complex programming like other titration systems on the market. This system offers superb dependability so your lab doesn’t suffer from downtime and it’s easy to use for all skill levels, reducing operator time and training. In many cases the autotitrator will allow an operator to prepare the next sample while the instrument is performing the current measurement, thus improving lab throughput. The Hach AT1222 automatic titrator eliminates manual processes and operator interpretation, significantly improving accuracy and reliability so you can trust your measurements from day to day.
Catalog No. 50-199-7529
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.