Learn More
Andwin Scientific TITERBLOCK DEEP 96-WELL PLATE
SDP
Supplier:  Andwin Scientific 75569600
Encompass_Preferred
TiterBlock;deep 96-well plates, polypropylene, 2.0 ml capacity square wells, 20 plates per pack.
Catalog No. 50-820-889
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.