Learn More
Andwin Scientific TITER-TOPS 100/PK
SDP
Supplier:  Andwin Scientific 7053810
Encompass_Preferred
Made with 4.4 mil polyethylene film with an FDA approved non cytotoxic acrylic adhesive, Titer-Tops is a product that is ideal for preventing the evaporation of samples, the contamination of well to well, and the spillage of samples. The film is pre-cut in 6" x 3.25" rectangular shapes with a paper liner. This product is able to withstand chemicals and thermal conditions. Its adhesive is waterproof, solvent resistant, and suitable with all plastic microtiter plates. This is also UV transparent to 250nm. It is sold in a pack of 100.
Catalog No. 50-930-380
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.