Learn More
Andwin Scientific TISSUETEK U-CASSETTE LIL 500CS
SDP
Supplier:  Andwin Scientific 25608774
Encompass
The Tissue-Tek Uni-Cassette has a smooth, single action, fracture-back hinge that allows the cassette to fracture when closed, and then break smoothly with no difficulty. This product features a unique design of raised spacer nubs in order to keep the space between the cassettes for better exchange of processing reagents. The cassette also has a wider, thicker finger tab that not only lessens the strain on operators finger, but also makes the opening of the lid less complicated. It also features an oversize writing surface that permits easy organization and identification. This product is sold in the color of lilac.
Catalog No. NC9970303
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.