Learn More
Andwin Scientific TISSUE-TEK UNI-CASSETTE PINK
Supplier: Andwin Scientific 62500P
The Tissue-Tek® Uni-Cassette® has a smooth, single action, fracture-back hinge that allows the cassette to fracture when closed, and then break smoothly with no difficulty. This product features a unique design of raised spacer nubs in order to keep the space between the cassettes for better exchange of processing reagents. The cassette also has a wider, thicker finger tab that not only lessens the strain on operator’s finger, but also makes the opening of the lid less complicated. It also features an oversize writing surface that permits easy organization and identification. This product is sold in the color of pink.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.