Learn More
Andwin Scientific TISSUE TEK SAKURA 1 GAL CS4
SDP
Supplier:  Andwin Scientific 5990
Encompass_Preferred
The Tissue-Tek VIP Fixative removes the necessity for routine warm water flushing. It does not generate any precipitate in the presence of alcohol, contrary to all other 10% neutral buffered formalin products. It also does not form salts within the tissue processor lines which can result to expensive damages to any brands of tissues processors unless regular maintenance is performed. It is sold in the quantity of 1 gallon.
Catalog No. 50-899-90476
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.