Learn More
Andwin Scientific TISSUE TEK FILING CABINET
SDP
Supplier:  Andwin Scientific 25608866
Encompass
The Tissue-Tek� Embedding Ring Filing Cabinet offers permanent secure storage/reference for blocks embedded in Tissue-Tek� embedding rings. This product can hold 1000 rings. Its one-piece design removes the need of hinges and doors, which then permits dust-free storage.
Catalog No. NC9532780
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.