Learn More
Andwin Scientific Microtome Blade Accu-Edge® High Profile, PTFE Coated Stainless Steel, Ultra Sharp Edge, 14 mm Height, Disposable
Supplier: Andwin Scientific 4685
The Accu-Edge disposable high-profile blades by Sakura Finetek are compatible with all Accu-Edge blade holders that are specially designed for disposable blades 14 millimeters in height. This product is ideal for microtomy, cryotomy, and sectioning specimens without striations, distortion, or chattering. It is sold in dispensers that can store 50 blades and distribute them in a few seconds. The previously used blade is then automatically ejected.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.