Learn More
Electron Microscopy Sciences Tissue Matrices, 25x25 mm, Rectangular
Supplier:  Electron Microscopy Sciences 69013
Encompass_Preferred
The Tissue Matrix is designed to aid the basic research scientist in the free-hand dissection of a wide variety of different tissues and repeatedly slice fresh tissue at intervals as small as 1 mm. The TM's are constructed from high grade aluminum to allow for rapid temperature equilibration, ease of cleaning or sterilization and withstand the rigors of daily use. Channels at 1 mm intervals. Channel width is 0.3 mm.
Size: 25x25 mm
Type: Rectangular
Catalog No. 50-293-55
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.