Learn More
Tisch Scientific PTFE (Polytetrafluoroethylene) membrane rolls
Supplier:  Tisch Scientific RS40213
Encompass_Preferred
Tisch Scientific PTFE (Polytetrafluoroethylene) membrane rolls are highly stable, chemically inert and naturally hydrophobic proving them to be suitable for HPLC analysis.
Catalog No. 50-225-1489
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.