Learn More
Andwin Scientific TIS-TEK CASS W/LID WH 1500/CS
SDP
Supplier:  Andwin Scientific 417002
Encompass
The Tissue-Tek® Uni-Cassette® with lid has a smooth, single action, fracture-back hinge that allows the cassette to fracture when closed, and then break smoothly with no difficulty. This product features a unique design of raised spacer nubs in order to keep the space between the cassettes for better exchange of processing reagents. The cassette also has a wider, thicker finger tab that not only lessens the strain on operator’s finger, but also makes the opening of the lid less complicated. It also features an oversize writing surface that permits easy organization and identification. This product is sold in the case of 1500.
Catalog No. NC9821136
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.