Learn More
Andwin Scientific TIPONE 0.1-10 5L NATURAL PIPE
SDP
Supplier:  Andwin Scientific 11113800
Encompass_Preferred
TipOne 0.1-10 ul natural pipet tips come in racks that have a one-piece base that resists tipping during its usage. The hinged lid reamins open for easy pipettor access. Both the lid and the base can withstand repeated autoclaving. This product is graduated at 2.5 and 10 l. This product is certified free of RNase, DNase, DNA and pyrogens. This product comes in 10 racks of 96 tips (960 total tips).
Catalog No. 50-819-558
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.